missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TESC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11125-1-AP-20UL
This item is not returnable.
View return policy
Description
Essential for the coupling of ERK cascade activation with the expression of ETS family genes in megakaryocytic differentiation.Specifications
| TESC | |
| Polyclonal | |
| Unconjugated | |
| TESC | |
| CHP3, TESC, tescalcin, TSC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 288689, 54997, 57816 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96BS2, Q9JKL5 | |
| Tesc | |
| TESC Fusion Protein Ag1596 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction