missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Tesaglitazar

Product Code. 18095442
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18095442 50 mg 50mg
18085432 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095442 missing translation for 'mfr' Tocris Bioscience™ missing translation for 'supplierNo' 3965/50
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PPARα/γ agonist

Dual-specificity PPARα/γ agonist (IC50 values are 0.35 and 3.8 μM for PPARγ and PPARα respectively). Prevents atherosclerosis progression in E3L.CETP transgenic mice. Also reduces insulin resistance in obese Zucker rats. Orally active.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 251565-85-2
Molecular Formula C20H24O7S

Specifications

Quantity 50 mg
Formula Weight 408.47
Purity >98%
Molecular Weight (g/mol) 408.47
Chemical Name or Material Tesaglitazar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.