missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Tesaglitazar

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Dual-specificity PPARα/γ agonist (IC50 values are 0.35 and 3.8 μM for PPARγ and PPARα respectively). Prevents atherosclerosis progression in E3L.CETP transgenic mice. Also reduces insulin resistance in obese Zucker rats. Orally active.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 408.47 |
| Purity | >98% |
| Molecular Weight (g/mol) | 408.47 |
| Chemical Name or Material | Tesaglitazar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction