missing translation for 'onlineSavingsMsg'
Learn More

TES sodium salt, Thermo Scientific Chemicals

Product Code. p-7042488
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 15435029

Brand: Thermo Scientific Alfa Aesar J62706.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A zwitterionic Good's Buffer

TES has potential for a variety of biological applications. In many culture media that require metal cations, many buffers cannot be used due to chelation or precipitation (citrate or phosphate, for example). TES was found to be beneficial in a study of succinate oxidation. TES is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9 where additional buffering capacity is required. It is also useful in cell culture media formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TES has potential for a variety of biological applications. In many culture media that require metal cations, many buffers cannot be used due to chelation or precipitation (citrate or phosphate, for example). TES was found to be beneficial in a study of succinate oxidation. TES is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9 where additional buffering capacity is required. It is also useful in cell culture media formulations.

Solubility
Soluble in water (350 g/L) at 20°C, and methanol(very faint turbidity).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Chemically stable, and stable to temperature and concentration changes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70331-82-7
Molecular Formula C6H14NNaO6S
Molecular Weight (g/mol) 251.23
MDL Number MFCD00065482
InChI Key GUXMFAHOYNRHBI-UHFFFAOYSA-M
Synonym 2-([Tris(hydroxymethyl)methyl]amino)ethane-1-sulfonic acid sodium salt
PubChem CID 23702152
IUPAC Name sodium 2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}ethane-1-sulfonate
SMILES [Na+].OCC(CO)(CO)NCCS([O-])(=O)=O

Specifications

Color White
Physical Form Powder
Quantity 25 g
Beilstein 1957061
Merck Index 14,9176
Solubility Information Soluble in water (350g/L) at 20°C,and methanol(very faint turbidity).
Formula Weight 251.23
Chemical Name or Material TES sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.