missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TES Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117724
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
TES maps to a common fragile site on chromosome 7q31.2 designated FRA7G and this gene is similar to mouse Testin, a testosterone-responsive gene encoding a Sertoli cell secretory protein containing three LIM domains. LIM domains are double zinc-finger motifs that mediate protein-protein interactions between transcription factors, cytoskeletal proteins and signaling proteins. Multiple protein isoforms are encoded by transcript variants of the TES gene and Cancer-associated chromosomal changes often involve regions containing fragile sites involving the TES gene.
Specifications
| TES | |
| Polyclonal | |
| Unconjugated | |
| TES | |
| D6Ertd352e; TES; Tes1; TES1/TES2; Tes2; TESS; TESS-2; testin; testin 2; testin LIM domain protein; testin2; testis derived transcript; testis derived transcript (3 LIM domains) | |
| Rabbit | |
| Protein G | |
| RUO | |
| 26136 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3.651 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UGI8 | |
| TES | |
| Recombinant Human Testin protein (130-260aa). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction