missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TES, 0.2M buffer soln., pH 7.5

Product Code. 15433959
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15433959 250 mL 250mL
15423959 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15433959 Supplier Thermo Scientific Chemicals Supplier No. J61350.AK
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70331-82-7 | C6H14NNaO6S | 251.23 g/mol

TES, 0.2M buffer soln., is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9, and is also useful for studying succinate oxidation. It is also widely used as a buffer in biological applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TES, 0.2M buffer soln., is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9, and is also useful for studying succinate oxidation. It is also widely used as a buffer in biological applications.

Solubility
Soluble in water (350 g/L) at 20°C, and methanol(very faint turbidity).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70331-82-7
Molecular Formula C6H14NNaO6S
Molecular Weight (g/mol) 251.23
MDL Number MFCD00065482
InChI Key GUXMFAHOYNRHBI-UHFFFAOYSA-M
PubChem CID 23702152
SMILES [Na+].OCC(CO)(CO)NCCS([O-])(=O)=O

Specifications

Concentration 0.2 M
pH 7.5
Physical Form Liquid
Quantity 250 mL
Solubility Information Soluble in water (350g/L) at 20°C,and methanol(very faint turbidity).
IUPAC Name sodium 2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}ethane-1-sulfonate
Chemical Name or Material TES buffer soln.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.