missing translation for 'onlineSavingsMsg'
Learn More

Terutroban, MedChemExpress

Product Code. p-200096759
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16437248

Brand: Medchem Express HY16991

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Terutroban is a thromboxane-prostaglandin receptor antagonist.

Terutroban is a thromboxane-prostaglandin receptor antagonist.

Specifications

Chemical Name or Material Terutroban
CAS 165538-40-9
Color White
Purity Grade Notes Research
Molecular Formula C20H22ClNO4S
Quantity 10 mg
Synonym S-18886
Solubility Information DMSO : ≥ 22 mg/mL (53.93 mM)
SMILES O=C(O)CCC1=C2CC[C@@H](NS(=O)(C3=CC=C(Cl)C=C3)=O)CC2=CC=C1C
Molecular Weight (g/mol) 407.91
Formula Weight 407.91
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.