missing translation for 'onlineSavingsMsg'
Learn More

Terutroban, MedChemExpress

Product Code. 16437248
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16437248 10 mg 10mg
16403267 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16437248 Supplier Medchem Express Supplier No. HY16991

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Terutroban is a thromboxane-prostaglandin receptor antagonist.

Terutroban is a thromboxane-prostaglandin receptor antagonist.

Specifications

Chemical Name or Material Terutroban
CAS 165538-40-9
Color White
Purity Grade Notes Research
Molecular Formula C20H22ClNO4S
Quantity 10 mg
Synonym S-18886
Solubility Information DMSO : ≥ 22 mg/mL (53.93 mM)
SMILES O=C(O)CCC1=C2CC[C@@H](NS(=O)(C3=CC=C(Cl)C=C3)=O)CC2=CC=C1C
Molecular Weight (g/mol) 407.91
Formula Weight 407.91
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.