Learn More
Bunzl Healthcare™ Terumo™ Eccentric Luer-Slip Syringe
Manufactured with polypropylene for superior thermal and chemical resistance. Bunzl™ Terumo™ Eccentric Luer-Slip Syringes have a unique bevel design to facilitate the penetration of the skin and minimize trauma.
Brand:  Bunzl Healthcare™ TUSS-10ES1
Product Code. 13167604
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Clear polypropylene barrel and plunger; double contact ring
  • Integrated plunger stop prevents accidental pull-out
  • Sterile, Latex-free
  • Bold, precise scale markings
  • Complies to ISO 7886 (DIS 7886)
  • Individually wrapped in tamper evident peel packs
  • Supplied without needles
  • Suitable for gases or liquids
  • Eccentric luer lock
For needles, please reference our disposable needles range, ISO 7886 (DIS 7886): 13127634, 13117614, 13144724, 13187434, 12349219, 13104354.
For research use only, unless otherwise indicated.