missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TERT Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586750
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
Telomerase is a ribonucleoprotein polymerase that maintains telomere ends by addition of the telomere repeat TTAGGG. The enzyme consists of a protein component with reverse transcriptase activity, encoded by this gene, and an RNA component which serves as a template for the telomere repeat. Telomerase expression plays a role in cellular senescence, as it is normally repressed in postnatal somatic cells resulting in progressive shortening of telomeres. Deregulation of telomerase expression in somatic cells may be involved in oncogenesis. Studies in mouse suggest that telomerase also participates in chromosomal repair, since de novo synthesis of telomere repeats may occur at double-stranded breaks. Alternatively spliced variants encoding different isoforms of telomerase reverse transcriptase (TERT) have been identified; the full-length sequence of some variants has not been determined. Alternative splicing at this locus is thought to be one mechanism of regulation of telomerase activity.
Specifications
| TERT | |
| Polyclonal | |
| Unconjugated | |
| TERT | |
| CMM9; DKCA2; DKCB4; EST2; HEST2; hTERT; hTRT; PFBMFT1; RGD:70494}; TCS1; Telomerase catalytic subunit; telomerase reverse transcriptase; telomerase reverse transcriptase beta; telomerase-associated protein 2; TERT; tert {ECO:0000312; TP2; TR; TRT | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 21752, 301965, 7015 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14746, O70372, Q673L6 | |
| TERT | |
| A synthetic peptide corresponding to residues in Human TERT. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction