missing translation for 'onlineSavingsMsg'
Learn More

tert-Pentylmagnesium chloride, 1M in 2-MeTHF

Product Code. 15470998
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470998 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15470998 Supplier Thermo Scientific Alfa Aesar Supplier No. H54824.AE
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 28276-08-6 | C5H11ClMg | 130.90 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Store at 4°C. Air and moisture sensitive.
TRUSTED_SUSTAINABILITY

Spezifikation

CAS 28276-08-6
Density 0.831
Molecular Formula C5H11ClMg
Linear Formula CH3CH2C(CH3)2MgCl
MDL Number MFCD00010356
Quantity 100 mL
UN Number UN3399
Sensitivity Air and moisture sensitive
Synonym 1,1-dimethylpropylmagnesium chloride, magnesium, chloro 1,1-dimethylpropyl, t-amylmagnesium chloride, tert-pentylmagnesium chloride, nwfdyjdcsihfab-uhfffaoysa-m, 1,1-dimethylpropyl magnesium chloride, magnesium,chloro 1,1-dimethylpropyl, chloro 2-methylbutan-2-yl magnesium, tert-pentylmagnesium chloride, 1m in 2-methf, 1,1-dimethylpropylmagnesium chloride solution, 1.0 m in diethyl ether
Solubility Information Reacts with water.
InChI Key NWFDYJDCSIHFAB-UHFFFAOYSA-M
SMILES CCC(C)(C)[Mg]Cl
IUPAC Name magnesium;2-methylbutane;chloride
Molecular Weight (g/mol) 130.90
PubChem CID 4056867
Formula Weight 130.9
Concentration or Composition (by Analyte or Components) 1M in 2-MeTHF
Chemical Name or Material tert-Pentylmagnesium chloride
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.