missing translation for 'onlineSavingsMsg'
Learn More

tert-Pentylbenzene, 97%

Product Code. 11382276
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382276 10 g 10g
11392276 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382276 Supplier Thermo Scientific Alfa Aesar Supplier No. L03731.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2049-95-8 | C11H16 | 148.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2049-95-8
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00015187
InChI Key QHTJSSMHBLGUHV-UHFFFAOYSA-N
Synonym tert-amylbenzene, tert-pentylbenzene, 2-methyl-2-phenylbutane, 1,1-dimethylpropyl benzene, benzene, 1,1-dimethylpropyl, benzene, tert-pentyl, 2-phenyl-2-methylbutane, t-amylbenzene, 2-methylbutan-2-yl benzene, tert-amyl benzene
PubChem CID 16295
IUPAC Name 2-methylbutan-2-ylbenzene
SMILES CCC(C)(C)C1=CC=CC=C1

Especificaciones

Density 0.87
Boiling Point 188°C to 191°C
Flash Point 65°C (149°F)
Refractive Index 1.495
Quantity 10 g
Beilstein 2039053
Formula Weight 148.25
Percent Purity 97%
Chemical Name or Material tert-Pentylbenzene

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.