missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butylsulfinamide, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | tert-Butylsulfinamide |
| CAS | 146374-27-8 |
| Molecular Formula | C4H11NOS |
| Quantity | 5 g |
| Synonym | (±)-tert-Butylsulfinamide, 1,1-Dimethylethylsulfinamide, 2-Methyl-2-propanesulfinamide, 2-Methylpropan-2-sulfinamide, tert-Butylsulfinamide, 2-Methyl-2-propanesulfinamide |
| InChI Formula | InChI=1S/C4H11NOS/c1-4(2,3)7(5)6/h5H2,1-3H3 |
| SMILES | CC(C)(C)S(=O)N |
| IUPAC Name | 2-methylpropane-2-sulfinamide |
| Molecular Weight (g/mol) | 121.2 |
| Formula Weight | 121.0561 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction