missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylsulfinamide, 98%

Product Code. 10491654 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10491654 5 g Glass bottle 5g
10244914 1 g Glass bottle 1g
10061074 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10491654 Supplier Thermo Scientific Acros Supplier No. 426830050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 146374-27-8 | C4H11NOS | 121.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146374-27-8
Molecular Formula C4H11NOS
Molecular Weight (g/mol) 121.20
MDL Number MFCD01863616,MFCD05861480
InChI Key CESUXLKAADQNTB-UHFFFAOYNA-N
Synonym 2-methyl-2-propanesulfinamide, tert-butanesulfinamide, t-butylsulfinamide, tert-butylsulfinamide, 1,1-dimethylethylsulfinamide, 2-propanesulfinamide, 2-methyl, 2-methyl-propane-2-sulfinic acid amide, tert-butylsulphinamide, unii-i85yc2za8o, i85yc2za8o
PubChem CID 3382465
IUPAC Name 2-methylpropane-2-sulfinamide
SMILES CC(C)(C)S(N)=O

Specifications

Melting Point 97.0°C to 101.0°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CS(O)NH2
Quantity 5 g
Merck Index 15,3607
Formula Weight 121.2
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material tert-Butylsulfinamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.