missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylmagnesium chloride, 1M in MeTHF

Product Code. 15458047
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15458047 100 mL 100mL
15468047 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458047 Supplier Thermo Scientific Alfa Aesar Supplier No. H51164.AE
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 677-22-5 | C4H9ClMg | 116.87 g/mol

tert-Butylmagnesium chloride is a Grignard reagent used in organic synthesis. It is also involved in copper-catalyzed cross-coupling reaction with primary-alkyl halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butylmagnesium chloride is a Grignard reagent used in organic synthesis. It is also involved in copper-catalyzed cross-coupling reaction with primary-alkyl halides.

Solubility
Miscible with alcohol and water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 677-22-5
Molecular Formula C4H9ClMg
Molecular Weight (g/mol) 116.87
MDL Number MFCD00000465
InChI Key ZDRJSYVHDMFHSC-UHFFFAOYSA-M
Synonym tert-butylmagnesium chloride, tert-butylmagnesiumchloride, t-butylmagnesium chloride, t-butyl magnesium chloride, tert-butyl chloro magnesium, magnesium, chloro 1,1-dimethylethyl, tert-butylmagnesium chloride solution, 1.0 m in thf, tbumgcl, tert-bumgcl, t-bumgcl
PubChem CID 2724198
IUPAC Name magnesium;2-methylpropane;chloride
SMILES CC(C)(C)[Mg]Cl

Specifications

Linear Formula (CH3)3CMgCl
Quantity 100 mL
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information Miscible with alcohol and water.
Formula Weight 116.87
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material tert-Butylmagnesium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.