missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyllithium, 18% solution in heptane, AcroSeal™, Thermo Scientific™

Product Code. 12133751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12133751 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12133751 Supplier Thermo Scientific Acros Supplier No. 445921000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 594-19-4, 142-82-5
Density 0.6900g/mL
Flash Point −4°C
Packaging AcroSeal™ Glass bottle
Molecular Formula C4H9Li
Linear Formula (CH3)CLi
MDL Number MFCD00008795
Quantity 100 mL
Synonym tert-butyllithium, t-butyllithium, tbuli, lithium, 1,1-dimethylethyl, t-buli, lithium 2-methylpropane, tert-butyllithium solution, tert butyllithium, t-butyllithiurn
InChI Key BKDLGMUIXWPYGD-UHFFFAOYSA-N
SMILES [Li]C(C)(C)C
IUPAC Name tert-butyllithium
Molecular Weight (g/mol) 64.06
PubChem CID 638178
Formula Weight 64.06
Chemical Name or Material tert-Butyllithium
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Identifiant produit
  • tert-Butyllithium, 1.6-3.2 M (15-30%) solution in Heptane
Mention d'avertissement
  • Danger
Catégorie de danger
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Pyrophoric liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which, in contact with water, emits flammable gases Category 1
Mention de danger
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.