missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylhydroquinone, 97%

Product Code. 10583121 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
2 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
250g
2kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10583121 500 g Plastic bottle 500g
10716551 5 g Glass Bottle 5g
10573121 250 g Plastic bottle 250g
10487170 2 kg Plastic bottle 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10583121 Supplier Thermo Scientific Acros Supplier No. 150825000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1948-33-0 | C10H14O2 | 166.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1948-33-0
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00002344
InChI Key BGNXCDMCOKJUMV-UHFFFAOYSA-N
Synonym tert-butylhydroquinone, tbhq, 2-tert-butylhydroquinone, t-butylhydroquinone, mtbhq, t-butyl hydroquinone, 2-tert-butyl-1,4-benzenediol, 2-t-butylhydroquinone, mono-tert-butylhydroquinone, sustane
PubChem CID 16043
ChEBI CHEBI:78886
IUPAC Name 2-tert-butylbenzene-1,4-diol
SMILES CC(C)(C)C1=C(C=CC(=C1)O)O

Specifications

Melting Point 125.0°C to 130.0°C
Color Black-Tan to White
Boiling Point 295.0°C
Flash Point 171°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula (CH3)3CC6H3(OH)2
Quantity 500 g
Beilstein 06, IV, 6013
Solubility Information Solubility in water: 1g/100mL (25 c). Other solubilities: soluble in acetone (190/100g at 25 c),,soluble in ethanol (120/100g at 25 c),,soluble in ethyl acetate (100g/100g at 25 c),,soluble in dmso,diethyl ether;,insoluble in benzene,toluene
Formula Weight 166.22
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material tert-Butylhydroquinone, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butylhydroquinone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.