missing translation for 'onlineSavingsMsg'
Learn More

(tert-Butyldimethylsiloxy)acetaldehyde, 90%

Product Code. 10091274 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10091274 5 g Glass bottle 5g
10255024 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10091274 Supplier Thermo Scientific Acros Supplier No. 397510050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102191-92-4 | C8H18O2Si | 174.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102191-92-4
Molecular Formula C8H18O2Si
Molecular Weight (g/mol) 174.32
MDL Number MFCD01321229
InChI Key MEBFFOKESLAUSJ-UHFFFAOYSA-N
Synonym tert-butyldimethylsilyloxy acetaldehyde, 2-tert-butyldimethylsilyl oxy acetaldehyde, 2-tert-butyldimethylsilyloxy acetaldehyde, tert-butyldimethylsiloxy acetaldehyde, t-butyldimethylsilyloxyacetaldehyde, 2-tert-butyl dimethyl silyl oxyacetaldehyde, acetaldehyde, 1,1-dimethylethyl dimethylsilyl oxy, pubchem20197
PubChem CID 4187788
SMILES CC(C)(C)[Si](C)(C)OCC=O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Gray to White
Density 0.9150g/mL
Boiling Point 156°C to 157°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CSi(CH3)2OCH2C(O)H
Refractive Index 1.4310 to 1.4330 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.915
IUPAC Name 2-[(tert-butyldimethylsilyl)oxy]acetaldehyde
Formula Weight 174.31
Percent Purity 90%
Physical Form Powder
Chemical Name or Material (tert-Butyldimethylsiloxy)acetaldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (tert-Butyldimethylsiloxy)acetaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.