missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldimethylsilane, 95%

Product Code. 11976151 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11976151 5 g 5g
11986151 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11976151 Supplier Thermo Scientific Acros Supplier No. 445710050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29681-57-0 | C6H16Si | 116.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29681-57-0
Molecular Formula C6H16Si
Molecular Weight (g/mol) 116.28
InChI Key ILMRJRBKQSSXGY-UHFFFAOYSA-N
Synonym tert-butyldimethylsilane, t-butyldimethylsilane, tert-butyldimethylsilyl, silane, 1,1-dimethylethyl dimethyl, t-butyldimethylsilyl, t-butyl-dimethylsilyl, t-butyl-dimethyl-silyl, tert-butyl dimethylsilane, tert-butyldimethylsilyl group
PubChem CID 9898815
IUPAC Name tert-butyl(dimethyl)silicon
SMILES CC(C)(C)[Si](C)C

Specifications

Density 0.7010g/mL
Boiling Point 81°C to 83°C
Flash Point -11°C
Assay Percent Range 94% min. (GC)
Quantity 5 g
Specific Gravity 0.701
Formula Weight 116.28
Percent Purity 95%
Chemical Name or Material tert-Butyldimethylsilane
compliance-icons compliance-icons
Product Identifier
  • tert-Butyldimethylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.