missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldimethylchlorosilane, 97%

Product Code. 11494364
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 11494364

missing translation for 'mfr': Thermo Scientific Alfa Aesar A13064.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 18162-48-6 | C6H15ClSi | 150.721 g/mol

tert-Butyldimethylchlorosilane is used to protect hydroxyl group in organic synthesis. It finds application in the synthesis of prostaglandin. It is also used as an auxiliary material for hypolipaemics such as lovastatin and simvastatin. It plays an important role in the preparation of isoxazolines N-oxides from alpha-bromonitroalkanes. It acts as a versatile protecting reagent for amines, amides and alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyldimethylchlorosilane is used to protect hydroxyl group in organic synthesis. It finds application in the synthesis of prostaglandin. It is also used as an auxiliary material for hypolipaemics such as lovastatin and simvastatin. It plays an important role in the preparation of isoxazolines N-oxides from alpha-bromonitroalkanes. It acts as a versatile protecting reagent for amines, amides and alcohols.

Solubility
Soluble in chloroform and ethyl acetate. Insoluble in water.

Notes
Moisture sensitive. Store in cool place. Incompatible with bases, metals, oxidizing agents and alcohols.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 18162-48-6
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.721
MDL Number MFCD00000501
InChI Key BCNZYOJHNLTNEZ-UHFFFAOYSA-N
Synonym tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride
PubChem CID 28928
ChEBI CHEBI:85071
IUPAC Name tert-butyl-chloro-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)Cl

Specifikationer

Boiling Point 124°C to 126°C
Melting Point 86°C to 89°C
Quantity 100 g
Linear Formula (CH3)3CSi(CH3)2Cl
UN Number UN2925
Beilstein 505999
Flash Point 22°C (72°F)
Solubility Information Soluble in chloroform and ethyl acetate. Insoluble in water.
Formula Weight 150.73
Percent Purity 97%
Odor Pungent
Sensitivity Moisture sensitive
Density 0.81
Chemical Name or Material tert-Butyldimethylchlorosilane
Visa mer Visa mindre
compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • tert-Butyldimethylchlorosilane
Signalord
  • Danger
Riskkategorier
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 A
Faroangivelser
  • H228-Flammable solid.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Kompletterande information
  • MIXTURE LIST-Contains : tert-Butyldimethylchlorosilane

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.