missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butylbenzene, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | tert-Butylbenzene |
| CAS | 98-06-6 |
| Molecular Formula | C10 H14 |
| Quantity | 5 g |
| Synonym | Benzene','tert-butyl- (8CI), (1,1-Dimethylethyl)benzene, 2-Methyl-2-phenylpropane, Dimethylethylbenzene, NSC 6557, Phenyltrimethylmethane, Trimethylphenylmethane, t-Butylbenzene, tert-Butylbenzene |
| InChI Formula | InChI=1S/C10H14/c1-10(2,3)9-7-5-4-6-8-9/h4-8H,1-3H3 |
| SMILES | CC(C)(C)c1ccccc1 |
| IUPAC Name | tert-butylbenzene |
| Molecular Weight (g/mol) | 134.22 |
| Formula Weight | 134.1096 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction