missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylbenzene, 99%

Product Code. 11469523
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469523 1000 mL 1000mL
11479523 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469523 Supplier Thermo Scientific Alfa Aesar Supplier No. A12378.AU
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-06-6 | C10H14 | 134.222 g/mol

tert-Butylbenzene is an useful intermediate for manufacturing agrochemicals and pharmaceuticals. tert-Butyl Toluene is used as a solvent for resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butylbenzene is an useful intermediate for manufacturing agrochemicals and pharmaceuticals. tert-Butyl Toluene is used as a solvent for resins.

Solubility
Insoluble in water(0.03g/100 mL at 20°C) .

Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-06-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00008816
InChI Key YTZKOQUCBOVLHL-UHFFFAOYSA-N
Synonym t-butylbenzene, benzene, 1,1-dimethylethyl, benzene, tert-butyl, 2-methyl-2-phenylpropane, phenyltrimethylmethane, pseudobutylbenzene, dimethylethylbenzene, trimethylphenylmethane, 1,1-dimethylethylbenzene, tert-butyl benzene
PubChem CID 7366
IUPAC Name tert-butylbenzene
SMILES CC(C)(C)C1=CC=CC=C1

Specifications

Melting Point -58°C
Density 0.867
Boiling Point 168°C to 169°C
Flash Point 34°C (93°F)
Odor Characteristic
Refractive Index 1.492
Quantity 1000 mL
UN Number UN2709
Beilstein 1421537
Merck Index 14,1551
Solubility Information Insoluble in water(0.03g/100 mL at 20°C) .
Formula Weight 134.22
Percent Purity 99%
Chemical Name or Material tert-Butylbenzene
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butylbenzene
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: tert-Butylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.