missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl6-bromo-4-oxo-3,4-dihydro-1'H-spiro[1,3-benzoxazine-2,4'-piperidine]-1'-, Thermo Scientific™

Product Code. 10317112
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10317112 1 g Amber Glass Bottle 1g
10451453 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10317112 Supplier Thermo Scientific Maybridge Supplier No. MO07057DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-05-4
Molecular Formula C17H21BrN2O4
Molecular Weight (g/mol) 397.269
MDL Number MFCD05865135
InChI Key FKBBJWUNJKLMBK-UHFFFAOYSA-N
Synonym tert-butyl 6-bromo-4-oxo-3,4-dihydro-1'h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, tert-butyl 6-bromo-4-oxo-3,4-dihydrospiro benzo e 1,3 oxazine-2,4'-piperidine-1'-carboxylate, 6-bromospiro 1,3-benzoxazine-2,4'-piperidine-4 3h-one, n-boc protected, tert-butyl 6-bromo-4-oxo-3h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, tert-butyl 6-bromo-4-oxospiro 3h-1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, 6-bromo-3,4-dihydro-4-oxo-spiro 2h-1,3-benzoxazine-2,4'-piperidin-1'-carboxylic acid tert-butyl ester, spiro 2h-1,3-benzoxazine-2,4'-piperidine-1'-carboxylicacid,6-bromo-3,4-dihydro-4-oxo-,1,1-dimethylethyl ester, tert-butyl 6-bromo-3,4-dihydro-4-oxo-1'h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate
PubChem CID 2794762
IUPAC Name tert-butyl 6-bromo-4-oxospiro[3H-1,3-benzoxazine-2,4'-piperidine]-1'-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC2(CC1)NC(=O)C3=C(O2)C=CC(=C3)Br

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 397.27
Percent Purity 95%
Chemical Name or Material tert-Butyl 6-bromo-4-oxo-3,4-dihydro-1';H-spiro[1,3-benzoxazine-2,4';-piperidine]-1';-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.