missing translation for 'onlineSavingsMsg'
Learn More

tert-butyl6-bromo-4-oxo-3,4-dihydro-1'H-spiro[1,3-benzoxazine-2,4'-piperidine]-1'-, Thermo Scientific™

£79.40 - £250.00

Chemical Identifiers

CAS 690632-05-4
Molecular Formula C17H21BrN2O4
Molecular Weight (g/mol) 397.269
MDL Number MFCD05865135
InChI Key FKBBJWUNJKLMBK-UHFFFAOYSA-N
Synonym tert-butyl 6-bromo-4-oxo-3,4-dihydro-1'h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, tert-butyl 6-bromo-4-oxo-3,4-dihydrospiro benzo e 1,3 oxazine-2,4'-piperidine-1'-carboxylate, 6-bromospiro 1,3-benzoxazine-2,4'-piperidine-4 3h-one, n-boc protected, tert-butyl 6-bromo-4-oxo-3h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, tert-butyl 6-bromo-4-oxospiro 3h-1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, 6-bromo-3,4-dihydro-4-oxo-spiro 2h-1,3-benzoxazine-2,4'-piperidin-1'-carboxylic acid tert-butyl ester, spiro 2h-1,3-benzoxazine-2,4'-piperidine-1'-carboxylicacid,6-bromo-3,4-dihydro-4-oxo-,1,1-dimethylethyl ester, tert-butyl 6-bromo-3,4-dihydro-4-oxo-1'h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate
PubChem CID 2794762
IUPAC Name tert-butyl 6-bromo-4-oxospiro[3H-1,3-benzoxazine-2,4'-piperidine]-1'-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC2(CC1)NC(=O)C3=C(O2)C=CC(=C3)Br
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10451453
View Documents Promotion Details
Thermo Scientific Maybridge
MO07057CB
250 mg Amber Glass Bottle
£79.40
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10317112
View Documents Promotion Details
Thermo Scientific Maybridge
MO07057DA
1 g Amber Glass Bottle
£250.00
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

690632-05-4
397.269
FKBBJWUNJKLMBK-UHFFFAOYSA-N
2794762
CC(C)(C)OC(=O)N1CCC2(CC1)NC(=O)C3=C(O2)C=CC(=C3)Br
C17H21BrN2O4
MFCD05865135
tert-butyl 6-bromo-4-oxo-3,4-dihydro-1'h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, tert-butyl 6-bromo-4-oxo-3,4-dihydrospiro benzo e 1,3 oxazine-2,4'-piperidine-1'-carboxylate, 6-bromospiro 1,3-benzoxazine-2,4'-piperidine-4 3h-one, n-boc protected, tert-butyl 6-bromo-4-oxo-3h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, tert-butyl 6-bromo-4-oxospiro 3h-1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, 6-bromo-3,4-dihydro-4-oxo-spiro 2h-1,3-benzoxazine-2,4'-piperidin-1'-carboxylic acid tert-butyl ester, spiro 2h-1,3-benzoxazine-2,4'-piperidine-1'-carboxylicacid,6-bromo-3,4-dihydro-4-oxo-,1,1-dimethylethyl ester, tert-butyl 6-bromo-3,4-dihydro-4-oxo-1'h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate
tert-butyl 6-bromo-4-oxospiro[3H-1,3-benzoxazine-2,4'-piperidine]-1'-carboxylate

Specifications

690632-05-4
95%
MFCD05865135
FKBBJWUNJKLMBK-UHFFFAOYSA-N
tert-butyl 6-bromo-4-oxospiro[3H-1,3-benzoxazine-2,4'-piperidine]-1'-carboxylate
2794762
95%
95.0
C17H21BrN2O4
tert-butyl 6-bromo-4-oxo-3,4-dihydro-1'h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, tert-butyl 6-bromo-4-oxo-3,4-dihydrospiro benzo e 1,3 oxazine-2,4'-piperidine-1'-carboxylate, 6-bromospiro 1,3-benzoxazine-2,4'-piperidine-4 3h-one, n-boc protected, tert-butyl 6-bromo-4-oxo-3h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, tert-butyl 6-bromo-4-oxospiro 3h-1,3-benzoxazine-2,4'-piperidine-1'-carboxylate, 6-bromo-3,4-dihydro-4-oxo-spiro 2h-1,3-benzoxazine-2,4'-piperidin-1'-carboxylic acid tert-butyl ester, spiro 2h-1,3-benzoxazine-2,4'-piperidine-1'-carboxylicacid,6-bromo-3,4-dihydro-4-oxo-,1,1-dimethylethyl ester, tert-butyl 6-bromo-3,4-dihydro-4-oxo-1'h-spiro 1,3-benzoxazine-2,4'-piperidine-1'-carboxylate
CC(C)(C)OC(=O)N1CCC2(CC1)NC(=O)C3=C(O2)C=CC(=C3)Br
397.269
397.27
tert-Butyl 6-bromo-4-oxo-3,4-dihydro-1';H-spiro[1,3-benzoxazine-2,4';-piperidine]-1';-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications