Learn More
Thermo Scientific Maybridge tert-Butyl tetrahydropyrazine-1(2H)-carboxylate, 10g, 97%
Brand: Thermo Scientific Maybridge AW01051EA
tert-Butyl tetrahydropyrazine-1(2H)-carboxylate, 10g, 97%
Code : 31
Additional Details : CAS Number : 57260-71-6 Weight : 0.06000kg Transport : UN number : 3259 Chem class : 8 Pack group : III