missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl (S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | tert-Butyl (S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate |
| CAS | 69610-40-8 |
| Molecular Formula | C10H19NO3 |
| Quantity | 10 g |
| Synonym | (2S)-2-(Hydroxymethyl)-1-pyrrolidinecarboxylic Acid 1,1-Dimethylethyl Ester, (2S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (2S)-2-Hydroxymethylpyrrolidine-1-carboxylic Acid tert-butyl Ester, (2S)-Hydroxymethylpyrrolidine-1-carboxylic Acid tert-butyl Ester, (2S)-Hydroxymethylpyrrolidine-1-carboxylic Acid tert-butyl Ester, (S)-(+)-N-Boc-prolinol, (S)-(-)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (S)-1-BOC-2-Pyrrolidinemethanol, (S)-tert-Butyl 2-(hydroxymethyl)pyrrolidine-1-carboxylate, 1,1-Dimethylethyl (2S)-2-(hydroxymethyl)-1-pyrrolidinecarboxylate, 1-BOC-2-(S)-Pyrrolidinemethanol, BOC-L-prolinol, Boc-(S)-(-)-2-pyrrolidinemethanol, N-Boc-L-prolinol, N-[(tert-Butoxy)carbonyl]-L-prolinol, N-tert-Butoxycarbonyl-(2S)-2-pyrrolidinemethanol, tert-Butyl (2S)-2-(Hydroxymethyl)-1-pyrrolidinecarboxylate, tert-Butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate |
| InChI Formula | InChI=1S/C10H19NO3/c1-10(2,3)14-9(13)11-6-4-5-8(11)7-12/h8,12H,4-7H2,1-3H3/t8-/m0/s1 |
| SMILES | CC(C)(C)OC(=O)N1CCC[C@H]1CO |
| IUPAC Name | tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate |
| Molecular Weight (g/mol) | 201.26 |
| Formula Weight | 201.1365 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction