missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (S)-2-aminobutyrate hydrochloride, 95%, Thermo Scientific™

Product Code. 15402509
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53956-05-1
Molecular Formula C8H18ClNO2
Molecular Weight (g/mol) 195.687
MDL Number MFCD00237697
InChI Key OEPKETQBSXWOBJ-RGMNGODLSA-N
Synonym h-abu-otbu.hcl, s-tert-butyl 2-aminobutanoate hydrochloride, h-2-abu-otbu.hcl, h-2-abu-otbu hcl, tert-butyl 2s-2-aminobutanoate hydrochloride, s-2-aminobutyric acid t-butyl ester hydrochloride, butanoic acid, 2-amino-, 1,1-dimethylethyl ester, hydrochloride, s, 2s-2-aminobutyric acid tert-butyl ester hydrochloride, h-abu-otbu hcl, h-2-abu-otbu hydrochloride
PubChem CID 53628861
IUPAC Name tert-butyl (2S)-2-aminobutanoate;hydrochloride
SMILES CCC(C(=O)OC(C)(C)C)N.Cl

Specifications

Quantity 1 g
Beilstein 6935251
Formula Weight 195.69
Percent Purity 95%
Physical Form Solid
Chemical Name or Material tert-Butyl (S)-2-aminobutyrate hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'