missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (S)-2-aminobutyrate hydrochloride, 95%, Thermo Scientific™

Product Code. 15422509
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15422509

Brand: Thermo Scientific Alfa Aesar H65284.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53956-05-1
Molecular Formula C8H18ClNO2
Molecular Weight (g/mol) 195.687
MDL Number MFCD00237697
InChI Key OEPKETQBSXWOBJ-RGMNGODLSA-N
Synonym h-abu-otbu.hcl, s-tert-butyl 2-aminobutanoate hydrochloride, h-2-abu-otbu.hcl, h-2-abu-otbu hcl, tert-butyl 2s-2-aminobutanoate hydrochloride, s-2-aminobutyric acid t-butyl ester hydrochloride, butanoic acid, 2-amino-, 1,1-dimethylethyl ester, hydrochloride, s, 2s-2-aminobutyric acid tert-butyl ester hydrochloride, h-abu-otbu hcl, h-2-abu-otbu hydrochloride
PubChem CID 53628861
IUPAC Name tert-butyl (2S)-2-aminobutanoate;hydrochloride
SMILES CCC(C(=O)OC(C)(C)C)N.Cl

Specifications

Quantity 25 g
Beilstein 6935251
Formula Weight 195.69
Percent Purity 95%
Physical Form Solid
Chemical Name or Material tert-Butyl (S)-2-aminobutyrate hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.