missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (S)-2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate, 98%, Thermo Scientific™

Product Code. 15435525
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435525 5 g 5g
15445525 25 g 25g
15455525 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15435525 Supplier Thermo Scientific Alfa Aesar Supplier No. H61655.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109010-60-8
Molecular Formula C16H22N2O3
Molecular Weight (g/mol) 290.363
MDL Number MFCD03426719
InChI Key QTEDVVHLTMELTB-LBPRGKRZSA-N
Synonym s-tert-butyl 2-3-amino-2-oxo-2,3,4,5-tetrahydro-1h-benzo b azepin-1-yl acetate, s-atba, unii-5wj869a7ay, benazepril related compound f, tert-butyl s-2-3-amino-2-oxo-2,3,4,5-tetrahydrobenzo b azepin-1-yl acetate, tert-butyl-3-amino-2,3,4,5-tetrahydro-2-oxo-1h-3s-benzazepine-1-acetate, s-t-butyl 2-3-amino-2-oxo-2,3,4,5-tetrahydro-1h-benzo b azepin-1-yl acetate, tert-butyl 2-3s-3-amino-2-oxo-4,5-dihydro-3h-1-benzazepin-1-yl acetate, 1,1-dimethylethyl 3s-3-amino-2-oxo-2,3,4,5-tetrahydro-1h-1-benzazepin-1-yl acetate, tert-butyl 2-3s-3-amino-2-oxo-2,3,4,5-tetrahydro-1h-1-benzazepin-1-yl acetate
PubChem CID 6350624
IUPAC Name tert-butyl 2-[(3S)-3-amino-2-oxo-4,5-dihydro-3H-1-benzazepin-1-yl]acetate
SMILES CC(C)(C)OC(=O)CN1C2=CC=CC=C2CCC(C1=O)N

Specifications

Color White
Quantity 5 g
Formula Weight 290.36
Percent Purity 98%
Physical Form Powder
Chemical Name or Material tert-Butyl (S)-2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.