missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (R)-(+)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 95%

Product Code. 10037642 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10037642 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10037642 Supplier Thermo Scientific Acros Supplier No. 370890010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95715-87-0 | C11H19NO4 | 229.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 95715-87-0
Color Colorless to Yellow
Density 1.0600g/mL
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C11H19NO4
Refractive Index 1.4450 to 1.447
MDL Number MFCD00674064
Quantity 1 g
Merck Index 15, 4404
Specific Gravity 1.06
Specific Rotation +85° to +95° (20°C, 589nm) (c=1, CHCl3)
Synonym r-tert-butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate, unii-upk623lv60, r-4-formyl-2,2-dimethyl-oxazolidine-3-carboxylic acid tert-butyl ester, tert-butyl 4r-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate, r-+-3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde, tert-butyl r-+-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 3-oxazolidinecarboxylic acid, 4-formyl-2,2-dimethyl-, 1,1-dimethylethyl ester, 4r, r-garner aldehyde, r-3-boc-4-formyl-2,2-dimethyl-1,3-oxazolidine, r---3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde
InChI Key PNJXYVJNOCLJLJ-QMMMGPOBSA-N
SMILES CC(C)(C)OC(=O)N1[C@H](COC1(C)C)C=O
IUPAC Name tert-butyl (4R)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
Molecular Weight (g/mol) 229.28
PubChem CID 178792
Formula Weight 229.28
Percent Purity 95%
Physical Form Oil
Chemical Name or Material tert-Butyl (R)-(+)-4-formyl-2, 2-dimethyl-3-oxazolidinecarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • tert-Butyl (R)-(+)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.