missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl Phenyl Carbonate, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | tert-Butyl Phenyl Carbonate |
| CAS | 6627-89-0 |
| Molecular Formula | C11H14O3 |
| Quantity | 25 g |
| Synonym | Carbonic Acid tert-Butyl Phenyl Ester, 1,1-Dimethylethyl Phenyl Carbonate, 1-tert-Butoxycarbonyloxybenzene, NSC 60248, Phenyl tert-Butyl Carbonate |
| InChI Formula | InChI=1S/C11H14O3/c1-11(2,3)14-10(12)13-9-7-5-4-6-8-9/h4-8H,1-3H3 |
| SMILES | CC(C)(C)OC(=O)Oc1ccccc1 |
| IUPAC Name | tert-butyl phenyl carbonate |
| Molecular Weight (g/mol) | 194.23 |
| Formula Weight | 194.0943 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction