missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl peroxybenzoate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
Specifications
Melting Point | 8.0°C |
Color | Yellow |
Density | 1.0400g/mL |
Boiling Point | 75.0°C to 76.0°C (0.2 mmHg) |
Flash Point | 93°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Plastic bottle |
Molecular Formula | C11H14O3 |
Linear Formula | C6H5CO2OC(CH3)3 |
Show More |
Safety and Handling
Product Identifier
- tert-Butyl peroxybenzoate
- Danger
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Organic peroxide Type C & D
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H315-Causes skin irritation.
- H332-Harmful if inhaled.
- H242-Heating may cause a fire.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'