missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl peroxyacetate, 50% solution in aromatic free mineral spirit

Product Code. 10758922 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10758922 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10758922 Supplier Thermo Scientific Acros Supplier No. 349862500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-71-1 | C6H12O3 | 132.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
TRUSTED_SUSTAINABILITY

Specifications

Color Undesignated
CAS 107-71-1, 64742-48-9
Molecular Formula C6H12O3
MDL Number MFCD00048240
Synonym tert-butyl peroxyacetate, t-butyl peracetate, tert-butyl peracetate, lupersol 70, t-butyl peroxyacetate, ethaneperoxoic acid, 1,1-dimethylethyl ester, trigonox f-c50, unii-fj3f3s50cs, peroxyacetic acid, tert-butyl ester, fj3f3s50cs
InChI Key SWAXTRYEYUTSAP-UHFFFAOYSA-N
SMILES CC(=O)OOC(C)(C)C
IUPAC Name tert-butyl ethaneperoxoate
Molecular Weight (g/mol) 132.159
PubChem CID 61019
Formula Weight 132.16
Percent Purity 49 to 51%
Packaging Plastic bottle
Quantity 250 mL
Physical Form Solution
Chemical Name or Material tert-Butyl peroxyacetate, 50% Solution In Aromatic Free Mineral Spirit
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butyl peroxyacetate, 50% solution in aromatic free mineral spirit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Organic peroxide Type C & D
  • Respiratory sensitiser Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H242-Heating may cause a fire.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.