missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl Perbenzoate, TRC
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Specifications
Specifications
| Chemical Name or Material | tert-Butyl Perbenzoate |
| CAS | 614-45-9 |
| Molecular Formula | C11H14O3 |
| Quantity | 100 mg |
| Synonym | Benzoyl tert-butyl peroxide, CP 02, CP 02 (catalyst), Chaloxyd TBPB, Chaloxyd, TBPB-HA-M 1, Esperox 10, Interox TBPB-HA-M 1, Kayabutyl B, LQ-TBPB, Link-Cup, TBPB, Luperox P, Luperox PXL, NSC 674, Norox TBPB, Perbutyl Z, TBPB, TBPB-HA-M 1, TBPB-HA-M 3, TC 5, TC 5 (vulcanizer), Trigonox 93, Trigonox C, Trigonox C 50D, V 73, t-Butyl peroxybenzoate, tert-Butyl benzoyl peroxide, tert-Butyl peroxybenzoate |
| InChI Formula | InChI=1S/C11H14O3/c1-11(2,3)14-13-10(12)9-7-5-4-6-8-9/h4-8H,1-3H3 |
| SMILES | CC(C)(C)OOC(=O)c1ccccc1 |
| IUPAC Name | tert-butyl benzenecarboperoxoate |
| Molecular Weight (g/mol) | 194.23 |
| Formula Weight | 194.0943 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction