missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Oxobutyrate, TRC

Product Code. 30342956
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342956 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342956 Supplier Toronto Research Chemicals Supplier No. TRCB693435100ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl Oxobutyrate
CAS 1694-31-1
Molecular Formula C8H14O3
Quantity 100 mL
Synonym tert-Butyl Ester Acetoacetic Acid, 1,1-Dimethylethyl Acetoacetate, 3-Oxobutanoic Acid 1,1-dimethylethyl Ester, 3-Oxobutyric Acid tert-Butyl Ester, NSC 42869, tert-Butyl 3-Oxobutanoate, tert-Butyl 3-Oxobutyrate, tert-Butyl Acetoacetate, tert-Butyl Acetylacetate, tert-Butyl Acetylacetonate
InChI Formula InChI=1S/C8H14O3/c1-6(9)5-7(10)11-8(2,3)4/h5H2,1-4H3
SMILES CC(=O)CC(=O)OC(C)(C)C
IUPAC Name tert-butyl 3-oxobutanoate
Molecular Weight (g/mol) 158.19
Formula Weight 158.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.