missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl nitrite, tech. 90%

Product Code. 11392897
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392897 100 g 100g
11382897 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392897 Supplier Thermo Scientific Alfa Aesar Supplier No. L16135.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 540-80-7 | C4H9NO2 | 103.121 g/mol

tert-Butyl nitrite is employed as a reagent for diazotization and nitrosation of alcohols, thiols, amines and cycloalkanes. It plays an important role in photocatalyzed conversion of aryl and heteroarylamines to selenides. It finds application in the preparation of aryl azides from aryl amines. It is also useful for radical multifunctionalization reactions of aliphatic alkenes and source of nitric oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl nitrite is employed as a reagent for diazotization and nitrosation of alcohols, thiols, amines and cycloalkanes. It plays an important role in photocatalyzed conversion of aryl and heteroarylamines to selenides. It finds application in the preparation of aryl azides from aryl amines. It is also useful for radical multifunctionalization reactions of aliphatic alkenes and source of nitric oxide.

Solubility
Miscible with diethyl ether, chloroform, alcohol and carbon disulfide. Slightly miscible with water. Immiscible with glycerol.

Notes
Light sensitive. Store in cool place. Incompatible with strong reducing agents, powdered metals and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-80-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00002055
InChI Key IOGXOCVLYRDXLW-UHFFFAOYSA-N
Synonym t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono
PubChem CID 10906
IUPAC Name tert-butyl nitrite
SMILES CC(C)(C)ON=O

Specifications

Density 0.867
Boiling Point 61°C to 63°C
Flash Point −23°C (−9°F)
Odor Sweet
Refractive Index 1.369
Quantity 100 g
UN Number UN2351
Beilstein 1209339
Merck Index 14,1583
Solubility Information Miscible with diethyl ether,chloroform,alcohol and carbon disulfide. Slightly miscible with water. Immiscible with glycerol.
Formula Weight 103.12
Percent Purity 90%
Grade Technical
Chemical Name or Material tert-Butyl nitrite
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl nitrite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: tert-Butyl nitrite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.