missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-(Diaminomethylidene)carbamate, TRC

Product Code. 30415019
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415019 500 mg 500mg
30423945 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30415019 Supplier Toronto Research Chemicals Supplier No. TRCT204583500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl N-(Diaminomethylidene)carbamate
CAS 219511-71-4
Molecular Formula C6H13N3O2
Quantity 500 mg
Synonym 1,1-Dimethylethyl N-(Aminoiminomethyl)carbamate, Boc-guanidine, N-Boc-guanidine, N-tert-Butoxycarbonylguanidine, tert-Butoxycarbonylguanidine
InChI Formula InChI=1S/C6H13N3O2/c1-6(2,3)11-5(10)9-4(7)8/h1-3H3,(H4,7,8,9,10)
SMILES CC(C)(C)OC(=O)NC(=N)N
IUPAC Name tert-butyl N-carbamimidoylcarbamate
Molecular Weight (g/mol) 159.19
Formula Weight 159.1008
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.