missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-allylcarbamate, 97%

Product Code. 11331929
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331929 50 g 50g
11321929 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331929 Supplier Thermo Scientific Alfa Aesar Supplier No. H26014.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78888-18-3 | C8H15NO2 | 157.213 g/mol

tert-Butyl N-allylcarbamate is used in the synthesis of isoxazolidines. Synthesis of 5-substituted thiazolidin-2-ones is from the reaction of xanthates and tert-butyl N-allylcarbamates .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl N-allylcarbamate is used in the synthesis of isoxazolidines. Synthesis of 5-substituted thiazolidin-2-ones is from the reaction of xanthates and tert-butyl N-allylcarbamates .

Solubility
Insoluble in water. Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78888-18-3
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00191870
InChI Key AWARHXCROCWEAK-UHFFFAOYSA-N
Synonym tert-butyl allylcarbamate, tert-butyl n-allylcarbamate, tert-butyl n-prop-2-en-1-yl carbamate, n-boc-allylamine, t-butyl n-allylcarbamate, carbamic acid, 2-propenyl-, 1,1-dimethylethyl ester, acmc-209pfu, allyl tert-butyl carbamate, tert-butyl-n-allylcarbamate, n-tertbutoxycarbonyl allyl amine
PubChem CID 4067274
IUPAC Name tert-butyl N-prop-2-enylcarbamate
SMILES CC(C)(C)OC(=O)NCC=C

Specifications

Melting Point 34°C to 39°C
Boiling Point 48°C to 50°C (1mmHg)
Linear Formula H2C=CHCH2NHCO2C(CH3)3
Solubility Information Insoluble in water. Soluble in methanol.
Density 0.938
Quantity 50 g
Flash Point 79°C (175°F)
Formula Weight 157.21
Percent Purity 97%
Chemical Name or Material tert-Butyl N-allylcarbamate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.