missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-(4-piperidinylmethyl)carbamate, 97%, Thermo Scientific™

Product Code. 10757495
Change view
Click to view available options
Quantity:
250 mg
1 g
25 g
Unit Size:
1g
250mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10757495 25 g 25g
10243432 250 mg 250mg
10744023 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10757495 Supplier Thermo Scientific Maybridge Supplier No. SPB08455EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 135632-53-0
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
InChI Key VHYXAWLOJGIJPC-UHFFFAOYSA-N
Synonym tert-butyl piperidin-4-ylmethyl carbamate, 4-boc-aminomethyl piperidine, 4-tert-butoxycarbonylaminomethyl piperidine, 4-n-boc-aminomethyl piperidine, 4-boc-aminomethyl-piperidine, tert-butyl n-piperidin-4-ylmethyl carbamate, tert-butyl piperidin-4-ylmethylcarbamate, boc-4-aminomethyl piperidine
PubChem CID 723429
IUPAC Name tert-butyl N-(piperidin-4-ylmethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC1CCNCC1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 214.31
Percent Purity 95%
Chemical Name or Material tert-Butyl N-(4-piperidinylmethyl)carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.