missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-(4-formylbenzyl)carbamate, 97%, Thermo Scientific™

Product Code. 10563003
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10563003

Brand: Thermo Scientific Maybridge MO07448DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 156866-52-3
Molecular Formula C13H17NO3
Molecular Weight (g/mol) 235.283
InChI Key HRJJBEIFHFVBRT-UHFFFAOYSA-N
Synonym tert-butyl 4-formylbenzylcarbamate, tert-butyl n-4-formylbenzyl carbamate, tert-butyl n-4-formylphenyl methyl carbamate, tert-butyln-4-formylbenzyl carbamate, tert-butyl 4-formylbenzyl carbamate, tert-butyln-4-formylphenyl methyl carbamate, n-boc-4-aminomethylbenzaldehyde, tert-butyl n-4 formylbenzyl carbamate, 4-aminomethyl benzaldehyde,n-boc protected, 4-formyl-benzyl-carbamic acid t-butyl ester
PubChem CID 2794832
IUPAC Name tert-butyl N-[(4-formylphenyl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC=C(C=C1)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 235.28
Percent Purity 97%
Chemical Name or Material tert-Butyl N-(4-formylbenzyl)carbamate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.