missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-(4-formylbenzyl)carbamate, 97%, Thermo Scientific™

Product Code. p-4510266
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10682015 250 mg Amber Glass Bottle 250mg
10563003 1 g Amber Glass Bottle 1g
10082504 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10682015 Supplier Thermo Scientific Maybridge Supplier No. MO07448CB
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 156866-52-3
Molecular Formula C13H17NO3
Molecular Weight (g/mol) 235.283
InChI Key HRJJBEIFHFVBRT-UHFFFAOYSA-N
Synonym tert-butyl 4-formylbenzylcarbamate, tert-butyl n-4-formylbenzyl carbamate, tert-butyl n-4-formylphenyl methyl carbamate, tert-butyln-4-formylbenzyl carbamate, tert-butyl 4-formylbenzyl carbamate, tert-butyln-4-formylphenyl methyl carbamate, n-boc-4-aminomethylbenzaldehyde, tert-butyl n-4 formylbenzyl carbamate, 4-aminomethyl benzaldehyde,n-boc protected, 4-formyl-benzyl-carbamic acid t-butyl ester
PubChem CID 2794832
IUPAC Name tert-butyl N-[(4-formylphenyl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC=C(C=C1)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 235.28
Percent Purity 97%
Chemical Name or Material tert-Butyl N-(4-formylbenzyl)carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.