missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-(3-formylbenzyl)carbamate, 90%, Thermo Scientific™

Product Code. 10265334
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265334 5 g Amber Glass Bottle 5g
10310893 250 mg Amber Glass Bottle 250mg
10503073 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10265334 Supplier Thermo Scientific Maybridge Supplier No. MO07494DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 170853-04-0
Molecular Formula C13H17NO3
Molecular Weight (g/mol) 235.283
InChI Key JORXNWZTJLYRQA-UHFFFAOYSA-N
PubChem CID 2794834
IUPAC Name tert-butyl N-[(3-formylphenyl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC=CC(=C1)C=O

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym tert-butyl 3-formylbenzylcarbamate, tert-butyl n-3-formylphenyl methyl carbamate, tert-butyl 3-formylbenzyl carbamate, 3-aminomethyl benzaldehyde, n-boc protected, 3-formylbenzylamine, n-boc protected, tert-butyl n-3 formylbenzyl carbamate, tert-butyl n-3-formylbenzyl carbamate, tert-butyl3-formylbenzylcarbamate, 3-formylbenzyl carbamic acid tert-butyl ester, 3-formyl-benzyl-carbamic acid tert-butyl ester
Formula Weight 235.28
Percent Purity 90%
Chemical Name or Material tert-Butyl 3-formylbenzylcarbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.