missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl N-(3-amino-3-thioxopropyl)carbamate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | (CH3)3COC(O)NH(CH2)2C(S)NH2 |
Quantity | 10 g |
Formula Weight | 204.29 |
Percent Purity | 97% |
Chemical Name or Material | Tert-butyl N-(3-amino-3-thioxopropyl)carbamate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'