missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-(3-amino-3-thioxopropyl)carbamate, 97%, Thermo Scientific™

Product Code. 10173764
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173764 5 g Amber Glass Bottle 5g
10437832 1 g Amber Glass Bottle 1g
11376955 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10173764 Supplier Thermo Scientific Maybridge Supplier No. SPB08402DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 77152-97-7
Molecular Formula C8H16N2O2S
Molecular Weight (g/mol) 204.288
MDL Number MFCD02180883
InChI Key OBDMXQCRRWGEQM-UHFFFAOYSA-N
Synonym tert-butyl n-3-amino-3-thioxopropyl carbamate, tert-butyl 3-amino-3-thioxopropylcarbamate, tert-butyl n-2-carbamothioylethyl carbamate, tert-butyl 3-amino-3-thioxopropyl carbamate, 3-tert-butoxycarbonyl amino propanethioamide, tert-butyl 3-amino-3-thioxoprop-1-yl carbamate, 2-thiocarbamoylethyl carbamic acid tert-butyl ester, carbamic acid,n-3-amino-3-thioxopropyl-, 1,1-dimethylethyl ester, tert-butyl 2-thiocarbamoylethylcarbamate, 3-tert-butoxycarbonylamino propanethioamide
PubChem CID 2735653
IUPAC Name tert-butyl N-(3-amino-3-sulfanylidenepropyl)carbamate
SMILES CC(C)(C)OC(=O)NCCC(=S)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)3COC(O)NH(CH2)2C(S)NH2
Quantity 5 g
Formula Weight 204.29
Percent Purity 97%
Chemical Name or Material Tert-butyl N-(3-amino-3-thioxopropyl)carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.