missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl malonate, 95%, Thermo Scientific Chemicals

Product Code. 15445898
Change view
Click to view available options
Quantity:
2 mL
10 mL
Unit Size:
10mL
2mL
Product Code. Quantity unitSize
15445898 2 mL 2mL
15455898 10 mL 10mL
2 options
This item is not returnable. View return policy

Product Code. 15445898

Brand: Thermo Scientific Alfa Aesar H59540.9W

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42726-73-8
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00042856
InChI Key XPSYZCWYRWHVCC-UHFFFAOYSA-N
Synonym tert-butyl methyl malonate, t-butyl methyl malonate, 1-tert-butyl 3-methyl propanedioate, acmc-20ajkv, tert-butyl methylmalote, pubchem20476, methyl tert-butyl malonate, ksc493c8h, tert-butyl methyl malonaoate
PubChem CID 2733872
IUPAC Name 3-O-tert-butyl 1-O-methyl propanedioate
SMILES CC(C)(C)OC(=O)CC(=O)OC

Specifications

Density 1.03
Boiling Point 80°C (11 mmHg)
Flash Point 79°C (174°F)
Linear Formula CH3OCOCH2COOC(CH3)3
Refractive Index 1.4155
Quantity 2 mL
Beilstein 1772136
Formula Weight 174.19
Percent Purity 95%
Chemical Name or Material tert-Butyl methyl malonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.