missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl Methyl Ether, TRC
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Specifications
Specifications
| Chemical Name or Material | Methyl tert-butyl ether |
| CAS | 1634-04-4 |
| Molecular Formula | C5 H12 O |
| Quantity | 50 mL |
| Synonym | Methyl-tert-Butyl Ether, Propane, 2-methoxy-2-methyl-, Ether, tert-butyl methyl (6CI,7CI,8CI), 2-Methoxy-2-methylpropane, 1,1-Dimethylethyl methyl ether, 2-Methyl-2-methoxypropane, Driveron, MTBE, Methyl 1,1-dimethylethyl ether, Methyl tert-butyl ether, Methyl tertiary butyl ether, t-Butyl methyl ether, tert-Butoxymethane, tert-Butyl methyl ether |
| InChI Formula | InChI=1S/C5H12O/c1-5(2,3)6-4/h1-4H3 |
| SMILES | COC(C)(C)C |
| IUPAC Name | 2-methoxy-2-methylpropane |
| Molecular Weight (g/mol) | 88.15 |
| Formula Weight | 88.0888 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction