missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, HPLC Grade, 99+%

Product Code. 11378448
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
Unit Size:
1 L
16 L
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378448 4 x 4 L 16 L
11308458 1 L 1 L
11388448 4 x 1 L 4 L
11398448 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11378448 Supplier Thermo Scientific Alfa Aesar Supplier No. 040477.M6
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chromatography solvent | CAS: 1634-04-4 | C5H12O | 88.15 g/mol

Used as a Chromatography solvent. Acts as a fuel component in fuel for gasoline engines. They raise the oxygen content of gasoline(oxygenates).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Melting Point -109°C
Density 0.742 g/mL
Quantity 4 x 4 L
Flash Point -25°C (-13°F)
Odor Mild
Physical Form Liquid
Refractive Index 1.369
UN Number UN2350
Beilstein 1730942
Merck Index 14,6032
Boiling Point 55°C to 56°C
Solubility Information Slightly soluble in water. Highly soluble in ethanol and ethyl ether.
Formula Weight 88.15
Percent Purity ≥99%
Grade HPLC
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.