missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, ACS Reagent, ≥99%, Honeywell™ Riedel-de Haën™

Product Code. 15675550
Change view
Click to view available options
Quantity:
500 mL
1 L
2 L
2.5 L
25 L
4 x 2.5 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
2 L
2.5 L
25 L
500mL
Pack of 4
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15675550 500 mL Glass Bottle 500mL
15625550 1 L Glass Bottle 1 L
15655550 2 L Glass Bottle 2 L
15635550 2.5 L Glass Bottle 2.5 L
15665550 4 x 2.5 L Glass Bottle Pack of 4
15645550 25 L Stainless Steel Can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15675550 Supplier Honeywell-Riedel-de Haen Supplier No. 443808500ML
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Melting Point -109°C
Density 0.74g/mL at 25°C
Quantity 500 mL
Explosion Limit 0.151
Flash Point −33°C (−27.4°F)
Ignition Point 705°F
Odor Aromatic
Packaging Glass Bottle
Physical Form Liquid
Linear Formula (CH3)3COCH3
UN Number UN2398
Beilstein 1730942
Boiling Point 55.3°C
Solubility Information In water, 42g/L at 20°C
Formula Weight 88.15g/mol
Percent Purity ≥99%
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.