missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl methyl ether, ACS Reagent, ≥99%, Honeywell™ Riedel-de Haën™
Click to view available options
Quantity:
500 mL
1 L
2 L
2.5 L
25 L
4 x 2.5 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Melting Point | -109°C |
Linear Formula | (CH3)3COCH3 |
UN Number | UN2398 |
Beilstein | 1730942 |
Solubility Information | In water, 42g/L at 20°C |
Formula Weight | 88.15g/mol |
Percent Purity | ≥99% |
Density | 0.74g/mL at 25°C |
Boiling Point | 55.3°C |
Quantity | 2.5 L |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'