missing translation for 'onlineSavingsMsg'
Learn More

Tert-Butyl Methyl Ether, ≥99%, Honeywell™ Riedel-de-Haën™ CHROMASOLV™

Product Code. 15681740
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15681740 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15681740 Supplier Honeywell-Riedel-de Haen Supplier No. 202571L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CHROMASOLV™, for pesticide residue analysis,≥99.0%

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY

Specifications

CAS 1634-04-4
Density 0.74g/mL at 25°C
Quantity 1 L
Explosion Limit 0.151
Flash Point −33°C (−27.4°F)
Ignition Point 705°F
Packaging Glass Bottle
Molecular Formula C5H12O
Linear Formula (CH3)3COCH3
MDL Number MFCD00008812
UN Number UN2398
Beilstein 1730942
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
SMILES CC(C)(C)OC
IUPAC Name 2-methoxy-2-methylpropane
Molecular Weight (g/mol) 88.15
PubChem CID 15413
ChEBI CHEBI:27642
Formula Weight 88.15g/mol
Percent Purity ≥99.0%
Grade CHROMASOLV
Chemical Name or Material Tert-Butyl Methyl Ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.